Learn More
(4-Morpholin-4-yl-phenyl)-methanol, 95+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | [4-(morpholin-4-yl)phenyl]methanol |
Formula Weight | 193.24 |
Percent Purity | 95% |
Chemical Name or Material | (4-Morpholin-4-yl-phenyl)-methanol |