missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Methylumbelliferyl Alpha-L-Idopyranosiduronic Acid 2-Sulfate Disodium Salt, TRC

Artikelnummer. 30481126
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481126 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30481126 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3347151MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Methylumbelliferyl Alpha-L-Idopyranosiduronic Acid 2-Sulfate Disodium Salt
CAS 2181774-46-7
Renhedsgrad noter HPLC
Molekylær formel C16H14Na2O12S
Mængde 1 mg
Synonym 4-Methylumbelliferyl ?-L-Iduronic Acid-2-sulfate Sodium Salt, 4-Methyl-2-oxo-2H-1-benzopyran-7-yl ?-L-Idopyranosiduronic Acid 2-(Hydrogen Sulfate)
InChI formel InChI=1S/C16H16O12S.2Na/c1-6-4-10(17)26-9-5-7(2-3-8(6)9)25-16-14(28-29(22,23)24)12(19)11(18)13(27-16)15(20)21;;/h2-5,11-14,16,18-19H,1H3,(H,20,21)(H,22,23,24);;/q;2*+1/p-2/t11-,12-,13+,14+,16+;;/m0../s1
SMIL CC(C1=CC=C2O[C@H]3[C@@H]([C@@H](O)[C@H](O)[C@H](C(O[Na])=O)O3)OS(=O)(O[Na])=O)=CC(OC1=C2)=O
IUPAC navn sodium (2R,3S,4S,5R,6S)-3,4-dihydroxy-6-((4-methyl-2-oxo-2H-chromen-7-yl)oxy)-5-(sulfonatooxy)tetrahydro-2H-pyran-2-carboxylate
Molekylvægt (g/mol) 476.32
Formel vægt 476
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.