Få mere at vide
4-Methylumbelliferyl 6-Sulfo-2-acetamido-2-deoxy-Beta-D-glucopyranoside Potassium Salt, TRC
Artikelnummer. 30012585
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012585 25 mg
30005727 250 mg
30007998 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012585 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33500025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 210357-38-3
Molekylær formel C18 H20 N O11 S . K
Molekylvægt (g/mol) 497.51
Synonym 2H-1-Benzopyran-2-one, 7-[[2-(acetylamino)-2-deoxy-6-O-sulfo-β-D-glucopyranosyl]oxy]-4-methyl-, monopotassium salt (9CI), 4-Methylumbelliferyl 6-sulfo-2-acetamido-2-deoxy-β-D-glucopyranoside potassium salt, 7-[[2-(Acetylamino)-2-deoxy-6-O-sulfo-β-D-glucopyranosyl]oxy]-4-methyl-2H-1-benzopyran-2-one potassium salt, 4-Methylumbelliferyl β-D-N-acetylglucosamine 6-sulfate potassium salt, 4-Methylumbelliferyl 2-acetamido-2-deoxy-β-D-glucopyranoside-6-sulfate potassium salt
IUPAC navn potassium;[(2R,3S,4R,5R,6S)-5-acetamido-3,4-dihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxan-2-yl]methyl sulfate
SMIL [K+].CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](COS(=O)(=O)[O-])O[C@H]1Oc2ccc3C(=CC(=O)Oc3c2)C
Tekniske data
Kemisk navn eller materiale 4-Methylumbelliferyl 6-Sulfo-2-acetamido-2-deoxy-beta-D-glucopyranoside Potassium Salt
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C18H21NO11S.K/c1-8-5-14(21)29-12-6-10(3-4-11(8)12)28-18-15(19-9(2)20)17(23)16(22)13(30-18)7-27-31(24,25)26;/h3-6,13,15-18,22-23H,7H2,1-2H3,(H,19,20)(H,24,25,26);/q;+1/p-1/t13-,15-,16-,17-,18-;/m1./s1
Formel vægt 497.0394
Procent renhed >95%