Få mere at vide
4-Methylumbelliferyl 2-Acetamido-2-deoxy-Beta-D-galactopyranoside 6-Sulfate Potassium Salt, TRC
Artikelnummer. 30468114
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30468114 5 mg
30477957 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30468114 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3335405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 383160-14-3
Molekylær formel C18H20KNO11S
Molekylvægt (g/mol) 497.51
Synonym 7-[[2-(Acetylamino)-2-deoxy-6-O-sulfo-beta-D-galactopyranosyl]oxy]-4-methyl-2H-1-benzopyran-2-one Monopotassium Salt
IUPAC navn potassium;[(2R,3R,4R,5R,6S)-5-acetamido-3,4-dihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxan-2-yl]methyl sulfate
SMIL [K+].CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](COS(=O)(=O)[O-])O[C@H]1Oc2ccc3C(=CC(=O)Oc3c2)C
Tekniske data
Kemisk navn eller materiale 4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-galactopyranoside 6-Sulfate Potassium Salt
Mængde 5 mg
InChI formel InChI=1S/C18H21NO11S.K/c1-8-5-14(21)29-12-6-10(3-4-11(8)12)28-18-15(19-9(2)20)17(23)16(22)13(30-18)7-27-31(24,25)26;/h3-6,13,15-18,22-23H,7H2,1-2H3,(H,19,20)(H,24,25,26);/q;+1/p-1/t13-,15-,16+,17-,18-;/m1./s1
Formel vægt 497.0394