missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl 2,3,6-Tri-O-benzoyl-Beta-D-galactopyranoside, TRC

Product Code. 30475672
Change view
Click to view available options
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30475672 50 mg
30472408 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30475672 Supplier Toronto Research Chemicals Supplier No. TRCM33761050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 849207-61-0
Molekylær formel C37 H30 O11
Molekylvægt (g/mol) 650.63
IUPAC navn [(2R,3S,4S,5R,6S)-4,5-dibenzoyloxy-3-hydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxan-2-yl]methyl benzoate
SMIL CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H](COC(=O)c4ccccc4)[C@H](O)[C@H](OC(=O)c5ccccc5)[C@H]3OC(=O)c6ccccc6)ccc12

Specifications

Kemisk navn eller materiale 4-Methylumbelliferyl 2,3,6-Tri-O-benzoyl-beta-D-galactopyranoside
Mængde 50 mg
InChI formel InChI=1S/C37H30O11/c1-22-19-30(38)45-28-20-26(17-18-27(22)28)44-37-33(48-36(42)25-15-9-4-10-16-25)32(47-35(41)24-13-7-3-8-14-24)31(39)29(46-37)21-43-34(40)23-11-5-2-6-12-23/h2-20,29,31-33,37,39H,21H2,1H3/t29-,31+,32+,33-,37-/m1/s1
Formel vægt 650.1788
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.