Få mere at vide
4-Methylumbelliferyl 2,3,6-Tri-O-benzoyl-Beta-D-galactopyranoside, TRC
Artikelnummer. 30472408
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30472408 5 mg
30475672 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30472408 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3376105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 849207-61-0
Molekylær formel C37 H30 O11
Molekylvægt (g/mol) 650.63
IUPAC navn [(2R,3S,4S,5R,6S)-4,5-dibenzoyloxy-3-hydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxan-2-yl]methyl benzoate
SMIL CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H](COC(=O)c4ccccc4)[C@H](O)[C@H](OC(=O)c5ccccc5)[C@H]3OC(=O)c6ccccc6)ccc12
Tekniske data
Kemisk navn eller materiale 4-Methylumbelliferyl 2,3,6-Tri-O-benzoyl-beta-D-galactopyranoside
Mængde 5 mg
InChI formel InChI=1S/C37H30O11/c1-22-19-30(38)45-28-20-26(17-18-27(22)28)44-37-33(48-36(42)25-15-9-4-10-16-25)32(47-35(41)24-13-7-3-8-14-24)31(39)29(46-37)21-43-34(40)23-11-5-2-6-12-23/h2-20,29,31-33,37,39H,21H2,1H3/t29-,31+,32+,33-,37-/m1/s1
Formel vægt 650.1788