missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Methylumbelliferone-13C4, TRC

Artikelnummer. 30464331
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30464331 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30464331 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33300225MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4-Methylumbelliferone-13C4
CAS 1569304-40-0
Renhedsgrad noter HPLC
Molekylær formel 13C4C6H8O3
Mængde 25 mg
Synonym 7-Hydroxy-4-methylcoumarin, 7-Hydroxy-4-methyl-2H-1-benzopyran-2-one-13C4, 7-Hydroxy-4-methyl-coumarin, 7-Hydroxy-4-methyl-2-chromenone-13C4, 7-Hydroxy-4-methyl-2-oxo-3-chromene, 7-Hydroxy-4-methyl-2H-1-benzopyran-2-one-13C4, NSC 19026, NSC 9408, Omega 127-13C4, Pilot 447-13C4, β-Methylumbelliferone-13C4
InChI formel InChI=1S/C10H8O3/c1-6-4-10(12)13-9-5-7(11)2-3-8(6)9/h2-5,11H,1H3/i1+1,4+1,6+1,10+1
SMIL [13CH3][13C]1=[13CH][13C](=O)Oc2cc(O)ccc12
IUPAC navn 7-hydroxy-4-(1^{13}C)methyl-chromen-2-one
Molekylvægt (g/mol) 180.14
Formel vægt 180.0608
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.