Få mere at vide
4-Methylpyridine-d7, CDN
Artikelnummer. 30321862
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321862 1 g
30316555 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321862 Leverandør CDN Leverandørnr. CDND52571G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Picoline-d7
CAS 29372-29-0
Molekylær formel C6 D7 N
Mængde 1 g
Synonym Pyridine-2,3,5,6-d4, 4-(methyl-d3)- (9 CI, ACI), 4-(Methyl-d3)pyridine-2,3,5,6-d4 (ACI), 4-Picoline-d7 (7 CI), 4-Picoline-α,α,α,2,3,5,6-d7 (8 CI), 4-Methylpyridine-d7, Perdeuterated 4-picoline, Perdeuterio-4-picoline, 4-Methylpyridine-D7
InChI formel InChI=1 S/C6H7N/c1-6-2-4-7-5-3-6/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D
SMIL [2 H]c1nc([2 H])c([2 H])c(c1[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,3,5,6-tetradeuterio-4-(trideuteriomethyl)pyridine
Molekylvægt (g/mol) 100.1696
Formel vægt 100.1018 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity