Få mere at vide
4-Methylpentanoic-d11 Acid, CDN
Artikelnummer. 30328633
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328633 1 g
30312631 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328633 Leverandør CDN Leverandørnr. CDND23691G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methylpentanoic Acid-D11
CAS 344298-98-2
Molekylær formel C6 D11 H O2
Mængde 1 g
Synonym Pentanoic-2,2,3,3,4,5,5,5-d8 acid, 4-(methyl-d3)- (9 CI), 4-(Methyl-d3)pentanoic-2,2,3,3,4,5,5,5-d8 acid, 4-Methylpentanoic-d11 Acid, 4-Methylpentanoic Acid-D11, 4-Methylvaleric Acid-d11
InChI formel InChI=1 S/C6H12O2/c1-5(2)3-4-6(7)8/h5H,3-4H2,1-2H3,(H,7,8)/i1D3,2D3,3D2,4D2,5 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O
IUPAC navn 2,2,3,3,4,5,5,5-octadeuterio-4-(trideuteriomethyl)pentanoic acid
Molekylvægt (g/mol) 127.2261
Formel vægt 127.1528 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity