missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 4-Methylhistamine dihydrochloride

Description
4-Methylhistamine dihydrochloride is a potent, high affinity H4 receptor agonist (Ki = 7 nM) that displays > 100-fold selectivity over other human histamine receptor subtypes.
Specifications
Specifications
| Kemisk navn eller materiale | 5-(2-Aminoethyl)-4-methylimidazole dihydrochloride |
| CAS | 36376-47-3 |
| Mængde | 10 mg |
| Opløselighed | Soluble to 50mM in water |
| Molekylær formel | C6H11N3.2HCl |
| Formel vægt | Observed MW: 198.09 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?