Få mere at vide
4-Methylcyclohexanecarboxylic Acid Methyl Ester, TRC
Artikelnummer. 30476151
Skift visning
Mængde:
1 g
2.5 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476151 5 g
30475854 1 g
30471135 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30476151 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2949805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51181-40-9
Molekylær formel C9H16O2
Molekylvægt (g/mol) 156.22
Synonym 4-Methyl-cyclohexanecarboxylic Acid Methyl Ester, Methyl 4-Methylcyclohexanecarboxylate, Cyclohexanecarboxylic acid, 4-methyl-, methyl ester (6CI, 7CI, 9CI, ACI), methyl 4-methylcyclohexane-1-carboxylate
IUPAC navn methyl 4-methylcyclohexane-1-carboxylate
SMIL O=C(OC)C1CCC(C)CC1
Tekniske data
Kemisk navn eller materiale 4-Methylcyclohexanecarboxylic Acid Methyl Ester
Mængde 5 g
InChI formel InChI=1S/C9H16O2/c1-7-3-5-8(6-4-7)9(10)11-2/h7-8H,3-6H2,1-2H3
Formel vægt 156.12