Få mere at vide
4-Methylcatechol-d8, CDN
Artikelnummer. 30314207
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314207 0.1 g 0.1g
30312704 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314207 Leverandør CDN Leverandørnr. CDND68100.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methylcatechol-d8
CAS 1219803-18-5
Molekylær formel C7 2H8 O2
Mængde 0.1 g
InChI formel InChI=1 S/C7H8O2/c1-5-2-3-6(8)7(9)4-5/h2-4,8-9 H,1H3/i1D3,2 D,3 D,4 D/hD2
SMIL [2 H]Oc1c([2 H])c([2 H])c(c([2 H])c1O[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,2,5-trideuterio-3,4-dideuteriooxy-6-(trideuteriomethyl)benzene
Molekylvægt (g/mol) 132.19
Formel vægt 132.1026 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity