Få mere at vide
4-Methylbenzylidene Camphor, TRC
Artikelnummer. 30133023
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133023 10 g
17973865 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133023 Leverandør Toronto Research Chemicals Leverandørnr. TRCM29050010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 36861-47-9
Molekylær formel C18 H22 O
Molekylvægt (g/mol) 254.37
Synonym Methyl Benzylidene Camphor, 3-(4'-Methyl)benzylidene-bornan-2-one, Bicyclo[2.2.1]heptan-2-one, 1,7,7-trimethyl-3-[(4-methylphenyl)methylene]-, (±)-, 1,7,7-Trimethyl-3-[(4-methylphenyl)methylene]bicyclo[2.2.1]heptan-2-one, 1,7,7-Trimethyl-3-(4-methylbenzylidene)bicyclo[2.2.1]heptan-2-one, 3-(4-Methylbenzylidene)-DL-camphor, 3-(4-Methylbenzylidene)-dl-camphor, 3-(4-Methylbenzylidene)bornan-2-one, 3-(4-Methylbenzylidene)camphor, 3-(4'-Methylbenzylidene)camphor, 3-(p-Methylbenzylidene)-DL-camphor, 3-(p-Methylbenzylidene)camphor, 4-MBC, 4-Methylbenzylidenecamphor, Enzacamene, Eusolex 6300, Methylbenzylidenecamphor, Neo Heliopan MBC, Parsol 500, Parsol 5000, Uvinul MBC 95, p-Methylbenzylidenecamphor
IUPAC navn 4,7,7-trimethyl-2-[(4-methylphenyl)methylidene]bicyclo[2.2.1]heptan-3-one
SMIL Cc1ccc(C=C2C3CCC(C)(C2=O)C3(C)C)cc1
Tekniske data
Kemisk navn eller materiale Methyl Benzylidene Camphor
Renhedsgrad noter HPLC
Mængde 10 g
InChI formel InChI=1S/C18H22O/c1-12-5-7-13(8-6-12)11-14-15-9-10-18(4,16(14)19)17(15,2)3/h5-8,11,15H,9-10H2,1-4H3
Formel vægt 254.1671
Procent renhed >95%