Få mere at vide
4-Methylbenzotriazole-d3, TRC
Artikelnummer. 30474442
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30474442 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30474442 Leverandør Toronto Research Chemicals Leverandørnr. TRCM28878210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methylbenzotriazole-d3
CAS 1219151-49-1
Renhedsgrad noter HPLC
Molekylær formel C7H4D3N3
Mængde 10 mg
Synonym 4-(Methyl-d3)-1H-benzotriazole, 7-(Methyl-d3)-1H-benzotriazole, 4-(Methyl-d3)-1,2,3-benzotriazole, 4-(Methyl-d3)-1H-benzo[d][1,2,3]triazole
InChI formel InChI=1S/C7H7N3/c1-5-3-2-4-6-7(5)9-10-8-6/h2-4H,1H3,(H,8,9,10)/i1D3
SMIL [2H]C([2H])([2H])C1=C2C(NN=N2)=CC=C1
Molekylvægt (g/mol) 136.17
Procent renhed >95