Få mere at vide
4-Methylamino-d3 Antipyrine, TRC
Artikelnummer. 30467114
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30467114 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467114 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2856021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methylamino-d3 Antipyrine
CAS 1246820-06-3
Renhedsgrad noter HPLC
Molekylær formel C12H12D3N3O
Mængde 1 mg
Synonym 1,5-Dimethyl-2-phenyl-4-(trideuteriomethylamino)pyrazol-3-one, 4-Methylaminophenazone-D3 (N4-methyl-D3), 4-Methylamino-d3 antipyrine, 4-[(Methyl-d3)amino]antipyrine, 1,2-Dihydro-1,5-dimethyl-4-[(methyl-d3)amino]-2-phenyl-3H-pyrazol-3-one, N-(Methyl-d3)-4-aminoantipyrine, N-(Methyl-d3)-4-aminophenazone, 4-(Methyl-d3)amino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one
InChI formel InChI=1S/C12H15N3O/c1-9-11(13-2)12(16)15(14(9)3)10-7-5-4-6-8-10/h4-8,13H,1-3H3/i2D3
SMIL [2H]C([2H])([2H])NC1=C(C)N(C)N(C1=O)c2ccccc2
Molekylvægt (g/mol) 220.29
Procent renhed >95