Få mere at vide
4-(Methylamino)antipyrine 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Methylamino antipyrine |
| CAS | 519-98-2 |
| Analyt- eller komponentnavne | 4-Methylamino antipyrine |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H15 N3 O |
| Koncentration | 100 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 1,2-Dihydro-1,5-dimethyl-4-(methylamino)-2-phenyl-3H-pyrazol-3-one, Antipyrine, 4-(methylamino)- (6CI,8CI), 1,2-Dihydro-1,5-dimethyl-4-(methylamino)-2-phenyl-3H-pyrazol-3-one, 4-(Methylamino)antipyrine, 4-Methylaminophenazone, 4-Monomethylaminoantipyrine, 4-Monomethylaminophenazone, Methylaminoantipyrine, Methylaminophenazone, Monomethylaminoantipyrine, N-Methyl-4-aminoantipyrine, N-Methyl-4-aminophenazone, N-Methylaminoantipyrine, N-Methylaminophenazone, Noramidopyrine, Noraminopyrine, Metamizole Sodium Monohydrate Imp. C (EP) |
| InChI formel | InChI=1S/C12H15N3O/c1-9-11(13-2)12(16)15(14(9)3)10-7-5-4-6-8-10/h4-8,13H,1-3H3 |
| SMIL | CNC1=C(C)N(C)N(C1=O)c2ccccc2 |
| Vis mere |