Få mere at vide
4-(Methylamino)-1-(3-pyridyl-d4)-1-butanone Dihydrochloride, TRC
Artikelnummer. 30473216
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30473216 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473216 Leverandør Toronto Research Chemicals Leverandørnr. TRCM28700350MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Methylamino)-1-(3-pyridyl-d4)-1-butanone Dihydrochloride
CAS 764661-23-6
Molekylær formel C10 2H4 H10 N2 O . 2 Cl H
Mængde 50 mg
Synonym 1-Butanone','4-(methylamino)-1-(3-pyridinyl-2,4,5,6-d4)-','dihydrochloride (9CI), 1-Butanone','4-(methylamino)-1-(3-pyridinyl-2,4,5,6-d4)-','hydrochloride (1:2) (ACI), Pseudooxynicotine D4 (Pyridinyl D4) Dihydrochloride
InChI formel InChI=1S/C10H14N2O.2ClH/c1-11-6-3-5-10(13)9-4-2-7-12-8-9;;/h2,4,7-8,11H,3,5-6H2,1H3;2*1H/i2D,4D,7D,8D;;
SMIL Cl.Cl.[2H]c1nc([2H])c(C(=O)CCCNC)c([2H])c1[2H]
IUPAC navn 4-(methylamino)-1-(2,4,5,6-tetradeuteriopyridin-3-yl)butan-1-one;dihydrochloride
Molekylvægt (g/mol) 255.18
Formel vægt 254.0891