Få mere at vide
4'-Methylacetophenone-d10, CDN
Artikelnummer. 30319001
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319001 0.5 g 0.5g
30315642 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319001 Leverandør CDN Leverandørnr. CDND54950.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4'-Methylacetophenone-D10
CAS 358730-83-3
Molekylær formel C9 D10 O
Mængde 0.5 g
Synonym Ethanone-2,2,2-d3, 1-[4-(methyl-d3)phenyl-2,3,5,6-d4]- (9 CI), 4'-(Methyl-d3)acetophenone-2,2,2,2',3',5',6'-d7, 1-[4-(Methyl-d3)phenyl-2,3,5,6-d4]ethanone-2,2,2-d3, 4'-Methylacetophenone-D10, 4'-Methylacetophenone-d10
InChI formel InChI=1 S/C9H10O/c1-7-3-5-9(6-4-7)8(2)10/h3-6 H,1-2H3/i1D3,2D3,3 D,4 D,5 D,6 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1C(=O)C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,2,2-trideuterio-1-[2,3,5,6-tetradeuterio-4-(trideuteriomethyl)phenyl]ethanone
Molekylvægt (g/mol) 144.2367
Formel vægt 144.1359 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity