Få mere at vide
4-Methyl Hippuric Acid-d7, TRC
Artikelnummer. 30465944
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30465944 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465944 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3119775MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methyl Hippuric Acid-d7
CAS 1216588-60-1
Molekylær formel C10 D7 H4 N O3
Mængde 5 mg
Synonym 2-[[2,3,5,6-Tetradeuterio-4-(trideuteriomethyl)benzoyl]amino]acetic acid, N-(4-Methylbenzoyl)glycine-D7 (methylbenzoyl-D7), N-(4-Methylbenzoyl)glycine-d7, 4-Methylhippuric acid-d7, 2-[[4-(Methyl-d3)benzoyl-2,3,5,6-d4]amino]acetic acid, p-Methylhippuric acid-d7, p-Toluric acid-d7, 4-Methyl hippuric acid-d7
InChI formel InChI=1S/C10H11NO3/c1-7-2-4-8(5-3-7)10(14)11-6-9(12)13/h2-5H,6H2,1H3,(H,11,14)(H,12,13)/i1D3,2D,3D,4D,5D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1C(=O)NCC(=O)O)C([2H])([2H])[2H]
IUPAC navn 2-[[2,3,5,6-tetradeuterio-4-(trideuteriomethyl)benzoyl]amino]acetic acid
Molekylvægt (g/mol) 200.242
Formel vægt 200.118