Få mere at vide
4-(Methyl-d3-nitrosamino)-1-(3-pyridyl)-1-butanone, TRC
Artikelnummer. 30472320
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30472320 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472320 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32575250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Methyl-d3-nitrosamino)-1-(3-pyridyl)-1-butanone
CAS 86270-92-0
Molekylær formel C10 2H3 H10 N3 O2
Mængde 50 mg
InChI formel InChI=1S/C10H13N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8H,3,5,7H2,1H3/i1D3
SMIL [2H]C([2H])([2H])N(CCCC(=O)c1cccnc1)N=O
IUPAC navn N-(4-oxo-4-pyridin-3-ylbutyl)-N-(trideuteriomethyl)nitrous amide
Molekylvægt (g/mol) 210.25
Formel vægt 210.1196