Få mere at vide
4-Methyl-d3-catechol, CDN
Artikelnummer. 30312575
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30312575 0.05 g -
30309115 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312575 Leverandør CDN Leverandørnr. CDND53980.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methylcatechol methyl-d3
CAS 83719-41-9
Molekylær formel C7 2H3 H5 O2
Mængde 0.05 g
InChI formel InChI=1 S/C7H8O2/c1-5-2-3-6(8)7(9)4-5/h2-4,8-9 H,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])c1ccc(O)c(O)c1
IUPAC navn 4-(trideuteriomethyl)benzene-1,2-diol
Molekylvægt (g/mol) 127.16
Formel vægt 127.0713 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity