Få mere at vide
4-(Methyl-d3-amino)-1-(3-pyridyl)-1-butanone Dihydrochloride, TRC
Artikelnummer. 30479884
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30479884 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30479884 Leverandør Toronto Research Chemicals Leverandørnr. TRCM28700210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Methyl-d3-amino)-1-(3-pyridyl)-1-butanone Dihydrochloride
CAS 1189727-40-9
Molekylær formel C10 2H3 H11 N2 O . 2 Cl H
Mængde 10 mg
InChI formel InChI=1S/C10H14N2O.2ClH/c1-11-6-3-5-10(13)9-4-2-7-12-8-9;;/h2,4,7-8,11H,3,5-6H2,1H3;2*1H/i1D3;;
SMIL Cl.Cl.[2H]C([2H])([2H])NCCCC(=O)c1cccnc1
IUPAC navn 1-pyridin-3-yl-4-(trideuteriomethylamino)butan-1-one;dihydrochloride
Molekylvægt (g/mol) 254.17
Formel vægt 253.0828