Artikelnummer. 30468271
Klik for at se tilgængelige muligheder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
Mængde
unitSize
30468271
2.5 mg
2.5mg
30465922
25 mg
-
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer.
30468271
Leverandør
Toronto Research Chemicals
Leverandørnr.
TRCM2943172.5MG
Please sign in to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Kemiske identifikatorer
CAS
1794940-24-1
Molekylær formel
C12H15D3ClNO
Molekylvægt (g/mol)
230.75
Synonym
2-[(Methyl-d3)amino]-1-(4-methylphenyl)-1-butanone Hydrochloride
IUPAC navn
1-(4-methylphenyl)-2-(trideuteriomethylamino)butan-1-one;hydrochloride
SMIL
O=C(C(NC([2H])([2H])[2H])CC)C1=CC=C(C)C=C1.Cl
Tekniske data
Kemisk navn eller materiale
4-Methyl Buphedrone-d3 Hydrochloride
Mængde
2.5 mg
InChI formel
InChI=1S/C12H17NO.ClH/c1-4-11(13-3)12(14)10-7-5-9(2)6-8-10;/h5-8,11,13H,4H2,1-3H3;1H/i3D3;
Formel vægt
230.1265
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores
Privatlivspolitik .
Ser du en mulighed for forbedring?