missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Methyl-3-thiosemicarbazide, TRC

Artikelnummer. 30482693
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 g
5 g
25 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30482693 25 g -
30467615 5 g -
30474180 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30482693 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22065325G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 6610-29-3
Molekylær formel C2H7N3S
Molekylvægt (g/mol) 105.16
Synonym Semicarbazide, 4-methyl-3-thio- (6CI,7CI,8CI), N-Methylhydrazinecarbothioamide, 1-Amino-3-methylthiourea, 3-Amino-1-methylthiourea, 4-Methyl-3-thiosemicarbazide, 4-Methylthiosemicarbazide, 4-Methylthiosemicarbazone, 4-N-Methylthiosemicarbazide, B 1130, Methylthiosemicarbazide, N-Methylthiosemicarbazide, NSC 56911, Semicarbazide, 4-methyl-3-thio- (6CI,7CI,8CI), N-Methylhydrazinecarbothioamide, 1-Amino-3-methylthiourea, 3-Amino-1-methylthiourea, 4-Methyl-3-thiosemicarbazide, 4-Methylthiosemicarbazide, 4-Methylthiosemicarbazone, 4-N-Methylthiosemicarbazide, B 1130, Methylthiosemicarbazide, N-Methylthiosemicarbazide, NSC 56911
IUPAC navn 1-amino-3-methylthiourea
SMIL CNC(=S)NN

Tekniske data

Kemisk navn eller materiale 4-Methyl-3-thiosemicarbazide
Mængde 25 g
InChI formel InChI=1S/C2H7N3S/c1-4-2(6)5-3/h3H2,1H3,(H2,4,5,6)
Formel vægt 105.0361
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.