Få mere at vide
4-Methyl-3-penten-2-one-d10, CDN
Artikelnummer. 30313707
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313707 1 g
30321275 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313707 Leverandør CDN Leverandørnr. CDND11171G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methylpent-3-en-2-one-D10
CAS 207456-83-5
Molekylær formel C6 D10 O
Mængde 1 g
Synonym 3-Penten-2-one-1,1,1,3,5,5,5-d7, 4-(methyl-d3)- (9 CI, ACI), 4-(Methyl-d3)-3-penten-2-one-1,1,1,3,5,5,5-d7 (ACI), 4-Methyl-3-penten-2-one-d10, 4-Methylpent-3-en-2-one-D10, Mesityl oxide-d10
InChI formel InChI=1 S/C6H10O/c1-5(2)4-6(3)7/h4H,1-3H3/i1D3,2D3,3D3,4 D
SMIL [2 H]C(=C(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C(=O)C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,3,5,5,5-heptadeuterio-4-(trideuteriomethyl)pent-3-en-2-one
Molekylvægt (g/mol) 108.2046
Formel vægt 108.1359 g/mol
Procent renhed 99 atom % D, min 96% Chemical Purity