Få mere at vide
4-Methyl-3-penten-2-ol, TRC
Artikelnummer. 30179542
Skift visning
Mængde:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179542 50 mg
30179368 25 mg
30179575 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179542 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22247850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4325-82-0
Molekylær formel C6H12O
Molekylvægt (g/mol) 100.15
IUPAC navn 4-methylpent-3-en-2-ol
SMIL CC(O)C=C(C)C
Tekniske data
Kemisk navn eller materiale 4-Methyl-3-penten-2-ol
Mængde 50 mg
InChI formel InChI=1S/C6H12O/c1-5(2)4-6(3)7/h4,6-7H,1-3H3
Formel vægt 100.0888