missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(tri-n-butylstannyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog Number p-7033997
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
301652-23-3
C18H33NSn
382.179
MFCD07787403
SHMPGQWVDZTZBX-UHFFFAOYSA-N
4-methyl-2-tributylstannyl pyridine, 4-methyl-2-tributylstannanyl pyridine, 4-methyl-2-tributylstannyl-pyridine, pyridine, 4-methyl-2-tributylstannyl, acmc-209hdi, 4-methyl-2-tributylstannanylpyridine, 4-methyl-2-tri-n-butylstannyl pyridine
11222775
tributyl-(4-methylpyridin-2-yl)stannane
CCCC[Sn](CCCC)(CCCC)C1=NC=CC(=C1)C
This item is not returnable. View return policy
301652-23-3
C18H33NSn
382.179
MFCD07787403
SHMPGQWVDZTZBX-UHFFFAOYSA-N
4-methyl-2-tributylstannyl pyridine, 4-methyl-2-tributylstannanyl pyridine, 4-methyl-2-tributylstannyl-pyridine, pyridine, 4-methyl-2-tributylstannyl, acmc-209hdi, 4-methyl-2-tributylstannanylpyridine, 4-methyl-2-tri-n-butylstannyl pyridine
11222775
tributyl-(4-methylpyridin-2-yl)stannane
CCCC[Sn](CCCC)(CCCC)C1=NC=CC(=C1)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2788
Formula Weight 382.16
Percent Purity 96%
Chemical Name or Material 4-Methyl-2-(tri-n-butylstannyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'