Få mere at vide
4-Methyl-2-nitroaniline-d6, TRC
Artikelnummer. 30464767
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30464767 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30464767 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32345210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methyl-2-nitroaniline-d6
CAS 1246820-08-5
Molekylær formel C7H2D6N2O2
Mængde 10 mg
Synonym 4-Methyl-2-nitro-benzenamine-d6, 2-Nitro-p-toluidine-d6, (4-Methyl-2-nitrophenyl)amine-d6, 1-Amino-2-nitro-4-methylbenzene-d6, 3-Nitro-4-aminotoluene-d6, 4-Amino-3-nitrotoluene-d6, Amarthol Fast Red GL Salt-d6, Azoamine Red A-d6, Mitsui Red GL Base-d6, NSC 2759-d6
InChI formel InChI=1S/C7H8N2O2/c1-5-2-3-6(8)7(4-5)9(10)11/h2-4H,8H2,1H3/i1D3,2D,3D,4D
SMIL [2H]c1c([2H])c(c([2H])c(c1N)[N+](=O)[O-])C([2H])([2H])[2H]
IUPAC navn 2,3,5-trideuterio-6-nitro-4-(trideuteriomethyl)aniline
Molekylvægt (g/mol) 158.19
Formel vægt 158.0962