Få mere at vide
(4-Methyl-1H-imidazol-5-yl)methanol Hydrochloride, TRC
Artikelnummer. 30466903
Skift visning
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30466903 2.5 g 2.5g
30480494 250 mg -
30482462 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30466903 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3135752.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38585-62-5
Molekylær formel C5 H8 N2 O . Cl H
Molekylvægt (g/mol) 148.59
Synonym 1H-Imidazole-5-methanol, 4-methyl-, hydrochloride (1:1), 1H-Imidazole-4-methanol, 5-methyl-, monohydrochloride (9CI), (4-Methyl-1H-imidazol-5-yl)methanol hydrochloride, (5-Methyl-1H-imidazol-4-yl)methanol hydrochloride, 4-(Hydroxymethyl)-5-methylimidazole hydrochloride, 4-Methyl-1H-imidazole-5-methanol hydrochloride, 4-Methyl-5-imidazolemethanol hydrochloride, 5-Methyl-4-imidazolemethanol hydrochloride
IUPAC navn (5-methyl-1H-imidazol-4-yl)methanol;hydrochloride
SMIL Cl.Cc1[nH]cnc1CO
Tekniske data
Kemisk navn eller materiale (5-Methyl-1H-imidazol-4-yl)methanol Hydrochloride
Mængde 2.5 g
InChI formel InChI=1S/C5H8N2O.ClH/c1-4-5(2-8)7-3-6-4;/h3,8H,2H2,1H3,(H,6,7);1H
Formel vægt 148.0403