Få mere at vide
4-Methyl-11-propyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one, Mikromol™
Artikelnummer. 30050716
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050716 25 mg
17970986 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050716 Leverandør Mikromol Leverandørnr. MM1146.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Methyl-11-propyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methyl-11-propyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one
CAS 287980-85-2
Analyt- eller komponentnavne 4-Methyl-11-propyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one
Forsendelsestilstand Room Temperature
Molekylær formel C15 H16 N4 O
Certificeringer/compliance ISO 17025
Synonym Nevirapine Anhydrous Imp. C (EP), Nevirapine USP Related Compound C, Nevirapine USP RC C, 4-Methyl-11-propyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one, Nevirapine Related Compound C, Nevirapine Impurity C
InChI formel InChI=1S/C15H16N4O/c1-3-9-19-13-11(5-4-7-16-13)15(20)18-12-10(2)6-8-17-14(12)19/h4-8H,3,9H2,1-2H3,(H,18,20)
SMIL CCCN1c2nccc(C)c2NC(=O)c3cccnc13
IUPAC navn 4-methyl-11-propyl-5H-dipyrido[2,3-b:3',2'-e][1,4]diazepin-6-one
Molekylvægt (g/mol) 268.31
Mængde 25 mg
Formel vægt 268.1324 g/mol