Få mere at vide
4-Methyl-1-naphthoic acid, 97%, Thermo Scientific™
Artikelnummer. 15487258
Skift visning
Mængde:
0.5 g
10 g
Pakningsstørrelse:
0.5g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15487258 0.5 g 0.5g
15497258 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 15487258 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H60765.02
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4488-40-8
Molekylær formel C12H10O2
Molekylvægt (g/mol) 186.21
MDL nummer MFCD00671557
InChI nøgle SIVYRLBDAPKADZ-UHFFFAOYSA-N
Synonym 4-methyl-1-naphthoic acid, 4-methyl-1-naphthalenecarboxylic acid, 4-methyl-1-napthoic acid, 1-naphthalenecarboxylic acid, 4-methyl, 4-methyl-naphthalene-1-carboxylic acid, 4-methylnaphthalenecarboxylic acid, 4-methylnaphthoic acid, acmc-1aehw, 4-methyl-1-naphthoicacid, 4-methyl-1-naphtholic acid
PubChem CID 3014285
IUPAC navn 4-methylnaphthalene-1-carboxylic acid
SMIL CC1=C2C=CC=CC2=C(C=C1)C(O)=O
Tekniske data
Kemisk navn eller materiale 4-Methyl-1-naphthoic acid
Smeltepunkt 179°C to 181°C
Mængde 0.5 g
Formel vægt 186.21
Procent renhed 97%