Få mere at vide
4-Methoxytoluene-Alpha,Alpha,Alpha-d3, TRC
Artikelnummer. 30413002
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30413002 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413002 Leverandør Toronto Research Chemicals Leverandørnr. TRCP27173210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methoxytoluene-Alpha,Alpha,Alpha-d3
CAS 14202-49-4
Molekylær formel C8 D3 H7 O
Mængde 10 mg
Synonym Benzene','1-methoxy-4-(methyl-d3)- (9CI','ACI), 1-Methoxy-4-(methyl-d3)benzene (ACI), Anisole','p-methyl-d3- (6CI','7CI','8CI), 4-Methoxytoluene-α,α,α-d3, p-Methyl-d3-anisole, 1-Methoxy-4-methylbenzene-d3, 1-Methoxy-4-methylbenzene-D3 (4-methyl-D3)
InChI formel InChI=1S/C8H10O/c1-7-3-5-8(9-2)6-4-7/h3-6H,1-2H3/i1D3
SMIL [2H]C([2H])([2H])c1ccc(OC)cc1
IUPAC navn 1-methoxy-4-(trideuteriomethyl)benzene
Molekylvægt (g/mol) 125.1829
Formel vægt 125.092