missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxysalicylic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-158684
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2237-36-7
C8H8O4
168.15
MFCD00002450
MRIXVKKOHPQOFK-UHFFFAOYSA-N
4-methoxysalicylic acid, 2-hydroxy-p-anisic acid, 2-hydroxy-4-methoxy-benzoic acid, benzoic acid, 2-hydroxy-4-methoxy, 2-hydroxy-4-methoxybenzoes, 4-methoxysalicylicacid, pubchem18895, 4-methoxy salicylic acid, 4-methoxy-salicylic acid, acmc-1cs6b
75231
2-hydroxy-4-methoxybenzoic acid
COC1=CC=C(C(O)=O)C(O)=C1
This item is not returnable. View return policy
2237-36-7
C8H8O4
168.15
MFCD00002450
MRIXVKKOHPQOFK-UHFFFAOYSA-N
4-methoxysalicylic acid, 2-hydroxy-p-anisic acid, 2-hydroxy-4-methoxy-benzoic acid, benzoic acid, 2-hydroxy-4-methoxy, 2-hydroxy-4-methoxybenzoes, 4-methoxysalicylicacid, pubchem18895, 4-methoxy salicylic acid, 4-methoxy-salicylic acid, acmc-1cs6b
75231
2-hydroxy-4-methoxybenzoic acid
COC1=CC=C(C(O)=O)C(O)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 157.0°C to 159.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3OC6H3(OH)CO2H
Quantity 10 g
Beilstein 10, 379
Formula Weight 168.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Methoxysalicylic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'