Få mere at vide
4-Methoxyresorcinol, TRC
Artikelnummer. 30481885
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481885 100 mg
30476736 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30481885 Leverandør Toronto Research Chemicals Leverandørnr. TRCM267605100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6100-60-3
Molekylær formel C7H8O3
Molekylvægt (g/mol) 140.14
Synonym 4-Methoxy-1,3-benzenediol, 4-Methoxy-resorcinol, 2,4-Dihydroxyanisole, 3-Hydroxy-4-methoxyphenol, 4-Methoxyresorcinol
IUPAC navn 4-methoxybenzene-1,3-diol
SMIL COc1ccc(O)cc1O
Tekniske data
Kemisk navn eller materiale 4-Methoxyresorcinol
Mængde 100 mg
InChI formel InChI=1S/C7H8O3/c1-10-7-3-2-5(8)4-6(7)9/h2-4,8-9H,1H3
Formel vægt 140.0473