Få mere at vide
4’-Methoxypropiophenone-d3, TRC
Artikelnummer. 30478417
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30478417 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30478417 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22622825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4'-Methoxypropiophenone-d3
CAS 89717-81-7
Molekylær formel C10 2H3 H9 O2
Mængde 25 mg
Synonym 1-Propanone-3,3,3-d3, 1-(4-methoxyphenyl)- (9CI), 3,3,3-Trideuterio-1-(4-methoxyphenyl)propan-1-one, 4'-Methoxypropiophenone-d3, 4'-Methoxypropiophenone-D3 (propanoyl-3,3,3-D3), 1-(4-Methoxyphenyl)-1-propanone-3,3,3-d3, 1-Methoxy-4-(propanoyl-3,3,3-d3)benzene
InChI formel InChI=1S/C10H12O2/c1-3-10(11)8-4-6-9(12-2)7-5-8/h4-7H,3H2,1-2H3/i1D3
SMIL [2H]C([2H])([2H])CC(=O)c1ccc(OC)cc1
IUPAC navn 3,3,3-trideuterio-1-(4-methoxyphenyl)propan-1-one
Molekylvægt (g/mol) 167.22
Formel vægt 167.1026