Få mere at vide
4-Methoxycarbonylmethyl-benzoic Acid Methyl Ester, TRC
Artikelnummer. 30471241
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30471241 250 mg
1 options
Artikelnummer. 30471241 Leverandør Toronto Research Chemicals Leverandørnr. TRCM329863250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methoxycarbonylmethyl-benzoic Acid Methyl Ester
CAS 52787-14-1
Molekylær formel C18 H35 N O2
Mængde 250 mg
Synonym Dimethyl Homoterephthalate, Dimethyl p-(Carboxymethyl)benzoate, Homoterephthalic Acid Dimethyl Ester, Methyl 4-(2-Methoxy-2-oxoethyl)benzoate, Methyl 4-Methoxycarbonylphenylacetate
InChI formel InChI=1S/C18H35NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h14-15,17,20H,2-13,16,19H2,1H3/b15-14+/t17-/m0/s1
SMIL CCCCCCCCCCCCC\C=C\C(=O)[C@@H](N)CO
IUPAC navn (E,2S)-2-amino-1-hydroxyoctadec-4-en-3-one
Molekylvægt (g/mol) 297.476
Formel vægt 297.267