Få mere at vide
4-Methoxybenzyl-d2 Alcohol, TRC
Artikelnummer. 30473047
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30473047 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473047 Leverandør Toronto Research Chemicals Leverandørnr. TRCM261069250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methoxybenzyl-d2 Alcohol
CAS 35693-15-3
Molekylær formel C8 D2 H8 O2
Mængde 250 mg
Synonym Benzenemethan-d2-ol, 4-methoxy-, 4-Methoxybenzenemethan-d2-ol, 4-Methoxy-[α-2H2]-benzyl alcohol, α,α-Dideutero-4-methoxybenzyl alcohol, 4-Methoxybenzenemethanol-D2 (methanol-D2), (4-Methoxyphenyl)methyl-d2 alcohol, 4-(Hydroxymethyl-d2)anisole, 4-Methoxybenzyl-α,α-d2 alcohol, Anise alcohol-d2, p-Anisyl alcohol-d2
InChI formel InChI=1S/C8H10O2/c1-10-8-4-2-7(6-9)3-5-8/h2-5,9H,6H2,1H3/i6D2
SMIL [2H]C([2H])(O)c1ccc(OC)cc1
IUPAC navn dideuterio-(4-methoxyphenyl)methanol
Molekylvægt (g/mol) 140.176
Formel vægt 140.081