Få mere at vide
4-Methoxybenzyl-alpha,alpha-d2 Alcohol, CDN
Artikelnummer. 30317916
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317916 0.25 g -
30313875 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317916 Leverandør CDN Leverandørnr. CDND75440.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methoxybenzyl-d2 Alcohol
CAS 35693-15-3
Molekylær formel C8 D2 H8 O2
Mængde 0.25 g
Synonym Benzenemethan-d2-ol, 4-methoxy-, 4-Methoxybenzenemethan-d2-ol, 4-Methoxy-[α-2H2]-benzyl alcohol, α,α-Dideutero-4-methoxybenzyl alcohol, 4-Methoxybenzenemethanol-D2 (methanol-D2), (4-Methoxyphenyl)methyl-d2 alcohol, 4-(Hydroxymethyl-d2)anisole, 4-Methoxybenzyl-α,α-d2 alcohol, Anise alcohol-d2, p-Anisyl alcohol-d2
InChI formel InChI=1 S/C8H10O2/c1-10-8-4-2-7(6-9)3-5-8/h2-5,9 H,6H2,1H3/i6D2
SMIL [2 H]C([2 H])(O)c1ccc(OC)cc1
IUPAC navn dideuterio-(4-methoxyphenyl)methanol
Molekylvægt (g/mol) 140.176
Formel vægt 140.081 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity