Få mere at vide
4-Methoxyaniline-2,3,5,6-d4, CDN
Artikelnummer. 30320438
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320438 0.25 g
30314121 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320438 Leverandør CDN Leverandørnr. CDND70110.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,3,5,6-Tetradeuterio-4-methoxyaniline
CAS 1219798-55-6
Molekylær formel C7 D4 H5 N O
Mængde 0.25 g
Synonym Benzen-2,3,5,6-d4-amine, 4-methoxy- (ACI), 4-Methoxybenzen-2,3,5,6-d4-amine (ACI), 2,3,5,6-Tetradeuterio-4-methoxyaniline, 4-Methoxyaniline-2,3,5,6-d4, 4-Methoxyaniline-2,3,5,6-D4, p-Anisidine-d4
InChI formel InChI=1 S/C7H9NO/c1-9-7-4-2-6(8)3-5-7/h2-5 H,8H2,1H3/i2D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c(OC)c([2 H])c([2 H])c1N
IUPAC navn 2,3,5,6-tetradeuterio-4-methoxyaniline
Molekylvægt (g/mol) 127.177
Formel vægt 127.094 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity