Få mere at vide
4-Methoxy Estrone-13C,d3, TRC
Artikelnummer. 30137681
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137681 10 mg
17908499 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30137681 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22613710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217437-34-7
Molekylær formel 13C C18 2H3 H21 O3
Molekylvægt (g/mol) 304.4
Synonym Estra-1,3,5(10)-trien-17-one, 3-hydroxy-4-(methoxy-13C-d3)-, 3-Hydroxy-4-methoxyestra-1,3,5(10)-trien-17-one-(methoxy-13C-D3), 4-Methoxyestrone-(methoxy-13C-D3)
IUPAC navn (8R,9S,13S,14S)-3-hydroxy-13-methyl-4-(trideuterio(1^{13}C)methoxy)-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one
SMIL [2H][13C]([2H])([2H])Oc1c(O)ccc2[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CCc12
Tekniske data
Kemisk navn eller materiale 4-Methoxyestrone methoxy-13C,d3
Mængde 10 mg
InChI formel InChI=1S/C19H24O3/c1-19-10-9-12-11-5-7-16(20)18(22-2)14(11)4-3-13(12)15(19)6-8-17(19)21/h5,7,12-13,15,20H,3-4,6,8-10H2,1-2H3/t12-,13-,15+,19+/m1/s1/i2+1D3
Formel vægt 304.1947