Få mere at vide
4-(Methoxy-d3)benzaldehyde, TRC
Artikelnummer. 30476865
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30476865 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476865 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33418710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Methoxy-d3)benzaldehyde
CAS 342611-04-5
Molekylær formel C8 D3 H5 O2
Mængde 10 mg
Synonym 4-Methoxybenzaldehyde-d3
InChI formel InChI=1S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3/i1D3
SMIL [2H]C([2H])([2H])Oc1ccc(C=O)cc1
IUPAC navn 4-(trideuteriomethoxy)benzaldehyde
Molekylvægt (g/mol) 139.17
Formel vægt 139.0713