Få mere at vide
4-Methoxy-d3 17beta-estradiol (>90%), TRC
Artikelnummer. 30482557
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30482557 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482557 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2626325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methoxy-d3-17beta-estradiol
CAS 1334143-12-2
Molekylær formel C19H23D3O3
Mængde 5 mg
Synonym Estra-1,3,5(10)-triene-3,17-diol, 4-(methoxy-d3)-, (17β)-, (17β)-4-(Methoxy-d3)estra-1,3,5(10)-triene-3,17-diol, 4-(Methoxy-d3)-17β-estradiol, 4-Methoxyestradiol-(methoxy-D3)
InChI formel InChI=1S/C19H26O3/c1-19-10-9-12-11-5-7-16(20)18(22-2)14(11)4-3-13(12)15(19)6-8-17(19)21/h5,7,12-13,15,17,20-21H,3-4,6,8-10H2,1-2H3
SMIL [2H]C([2H])([2H])Oc1c(O)ccc2[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CCc12
Molekylvægt (g/mol) 305.43