Få mere at vide
4-Methoxy-6-(2-phenylethyl)-2H-pyran-2-one, TRC
Artikelnummer. 30471951
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471951 1 mg
30470724 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30471951 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2645851MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3155-51-9
Molekylær formel C14H14O3
Molekylvægt (g/mol) 230.26
Synonym 5,6-Dehydro-7,8-dihydrokavain, 5,6-Didehydro-7,8-dihydrokawain, 5,6-Dehydro-7,8-dihydrokawain
SMIL O=C1C=C(OC)C=C(CCC2=CC=CC=C2)O1
Tekniske data
Kemisk navn eller materiale 4-Methoxy-6-(2-phenylethyl)-2H-pyran-2-one
Mængde 1 mg
InChI formel InChI=1S/C14H14O3/c1-16-13-9-12(17-14(15)10-13)8-7-11-5-3-2-4-6-11/h2-6,9-10H,7-8H2,1H3
Formel vægt 230.0943