missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methoxy-3-(trifluoromethoxy)benzonitrile, 97%

Brand:  Alfa Aesar™ H33310.MD

Additional Details : Weight : 0.00025kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 15438395

  • 310.00 DKK / 250mg

Estimated Shipment
17-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
4-Methoxy-3-(trifluoromethoxy)benzonitrile participate in the synthesis and structure-activity relationships of novel oxadiazole derivatives as GSK-3β inhibitors .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

1261653-03-5
217.147
DDKKATZYULLWQH-UHFFFAOYSA-N
66523525
COC1=C(C=C(C=C1)C#N)OC(F)(F)F
C9H6F3NO2
MFCD18398458
4-methoxy-3-trifluoromethoxy benzonitrile
4-methoxy-3-(trifluoromethoxy)benzonitrile

Specifications

4-Methoxy-3-(trifluoromethoxy)benzonitrile
1261653-03-5
MFCD18398458
4-methoxy-3-trifluoromethoxy benzonitrile
COC1=C(C=C(C=C1)C#N)OC(F)(F)F
217.147
217.14
250mg
C9H6F3NO2
UN3439
DDKKATZYULLWQH-UHFFFAOYSA-N
4-methoxy-3-(trifluoromethoxy)benzonitrile
66523525
97%
Videos
SDS Documents