missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-7032175
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
647855-22-9
C9H7F3O4
236.146
MFCD18398451
PHSHCIBRNMLSBJ-UHFFFAOYSA-N
4-methoxy-3-trifluoromethoxy benzoic acid, benzoic acid, 4-methoxy-3-trifluoromethoxy, 4-methoxy-3-trifluoromethoxy-benzoic acid
66523504
4-methoxy-3-(trifluoromethoxy)benzoic acid
COC1=C(C=C(C=C1)C(=O)O)OC(F)(F)F
This item is not returnable. View return policy
647855-22-9
C9H7F3O4
236.146
MFCD18398451
PHSHCIBRNMLSBJ-UHFFFAOYSA-N
4-methoxy-3-trifluoromethoxy benzoic acid, benzoic acid, 4-methoxy-3-trifluoromethoxy, 4-methoxy-3-trifluoromethoxy-benzoic acid
66523504
4-methoxy-3-(trifluoromethoxy)benzoic acid
COC1=C(C=C(C=C1)C(=O)O)OC(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 236.14
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethoxy)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'