missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-nitropyridine hydrochloride, 97%, Thermo Scientific™

Catalog Number p-7022368
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
31872-61-4
C6H7ClN2O3
190.583
MFCD00051995
RRJVDMBZMAGZGE-UHFFFAOYSA-N
4-methoxy-3-nitropyridine hydrochloride, 3-nitro-4-methoxypyridine hydrochloride, 4-methoxy-3-nitropyridine hydrogen chloride, pubchem6689, acmc-1ae1i, 4methoxy-3-nitropyridine hydrochloride, 4-methoxy-3-nitro-pyridine hydrochloride, pyridine, 4-methoxy-3-nitro-, monohydrochloride
355831
4-methoxy-3-nitropyridine;hydrochloride
COC1=C(C=NC=C1)[N+](=O)[O-].Cl
This item is not returnable. View return policy
31872-61-4
C6H7ClN2O3
190.583
MFCD00051995
RRJVDMBZMAGZGE-UHFFFAOYSA-N
4-methoxy-3-nitropyridine hydrochloride, 3-nitro-4-methoxypyridine hydrochloride, 4-methoxy-3-nitropyridine hydrogen chloride, pubchem6689, acmc-1ae1i, 4methoxy-3-nitropyridine hydrochloride, 4-methoxy-3-nitro-pyridine hydrochloride, pyridine, 4-methoxy-3-nitro-, monohydrochloride
355831
4-methoxy-3-nitropyridine;hydrochloride
COC1=C(C=NC=C1)[N+](=O)[O-].Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∼270°C (decomposition)
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 190.59
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-nitropyridine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'