Få mere at vide
4-Methoxy-2-nitroaniline-d3, TRC
Artikelnummer. 30471370
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30471370 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471370 Leverandør Toronto Research Chemicals Leverandørnr. TRCM264752500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Methoxy-d3)-2-nitroaniline
CAS 922730-95-8
Molekylær formel C7H5D3N2O3
Mængde 500 mg
InChI formel InChI=1S/C7H8N2O3/c1-12-5-2-3-6(8)7(4-5)9(10)11/h2-4H,8H2,1H3/i1D3
SMIL [2H]C([2H])([2H])Oc1ccc(N)c(c1)[N+](=O)[O-]
IUPAC navn 2-nitro-4-(trideuteriomethoxy)aniline
Molekylvægt (g/mol) 171.17
Formel vægt 171.0723