Få mere at vide
4-Methoxy-2-[[(5-methoxy-1H-benzimidazol-2-yl)sulphonyl]methyl]-3,5-dimethylpyridine 1-Oxide (Omeprazole Sulphone N-Oxide), Mikromol™
Beskrivelse
4-Methoxy-2-[[(5-methoxy-1H-benzimidazol-2-yl)sulphonyl]methyl]-3,5-dimethylpyridine 1-Oxide (Omeprazole Sulphone N-Oxide)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Omeprazole Sulfone N-Oxide |
| CAS | 158812-85-2 |
| Analyt- eller komponentnavne | Omeprazole Sulfone N-Oxide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H19 N3 O5 S |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Omeprazole Imp. I (EP), Omeprazole USP Related Compound I, Omeprazole USP RC I, 4-Methoxy-2-[[(5-methoxy-1H-benzimidazol-2-yl)sulphonyl]methyl]-3,5-dimethylpyridine 1-Oxide, Omeprazole Sulphone N-Oxide, Omeprazole Related Compound I, Omeprazole Impurity I |
| InChI formel | InChI=1S/C17H19N3O5S/c1-10-8-20(21)15(11(2)16(10)25-4)9-26(22,23)17-18-13-6-5-12(24-3)7-14(13)19-17/h5-8H,9H2,1-4H3,(H,18,19) |
| SMIL | COc1ccc2[nH]c(nc2c1)S(=O)(=O)Cc3c(C)c(OC)c(C)c[n+]3[O-] |
| IUPAC navn | 5-methoxy-2-[(4-methoxy-3,5-dimethyl-1-oxidopyridin-1-ium-2-yl)methylsulfonyl]-1H-benzimidazole |
| Vis mere |