Få mere at vide
4-Methoxy-2,3,6-trimethylbenzyl Alcohol-d3, TRC
Artikelnummer. 30472609
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30472609 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472609 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26980210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methoxy-2,3,6-trimethylbenzyl Alcohol-d3
CAS 1189501-77-6
Molekylær formel C11 2H3 H13 O2
Mængde 10 mg
InChI formel InChI=1S/C11H16O2/c1-7-5-11(13-4)9(3)8(2)10(7)6-12/h5,12H,6H2,1-4H3/i4D3
SMIL [2H]C([2H])([2H])Oc1cc(C)c(CO)c(C)c1C
IUPAC navn [2,3,6-trimethyl-4-(trideuteriomethoxy)phenyl]methanol
Molekylvægt (g/mol) 183.26
Formel vægt 183.1339