missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog Number p-3778901
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
536-60-7
C10H14O
150.22
MFCD00004663
OIGWAXDAPKFNCQ-UHFFFAOYSA-N
4-isopropylbenzyl alcohol, cuminol, cumic alcohol, cuminyl alcohol, cuminic alcohol, 4-isopropylphenyl methanol, cumyl alcohol, p-cymen-7-ol, cumin alcohol, p-isopropylbenzyl alcohol
325
CHEBI:27628
(4-propan-2-ylphenyl)methanol
CC(C)C1=CC=C(C=C1)CO
This item is not returnable. View return policy
536-60-7
C10H14O
150.22
MFCD00004663
OIGWAXDAPKFNCQ-UHFFFAOYSA-N
4-isopropylbenzyl alcohol, cuminol, cumic alcohol, cuminyl alcohol, cuminic alcohol, 4-isopropylphenyl methanol, cumyl alcohol, p-cymen-7-ol, cumin alcohol, p-isopropylbenzyl alcohol
325
CHEBI:27628
(4-propan-2-ylphenyl)methanol
CC(C)C1=CC=C(C=C1)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 28°C
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 248°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHC6H4CH2OH
Refractive Index 1.5180 to 1.52
Quantity 25 g
Beilstein 06, III, 1911
Merck Index 15, 2609
Specific Gravity 0.98
Formula Weight 150.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Isopropylbenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Isopropylbenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'